Assay Details
| Gene Symbol: ston2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033743.1 | NP_001028915.1 | 6 - 7 | 2621 | 113 | |
| XM_005169836.4 | XP_005169893.1 | 3 - 4 | 2236 | 113 | ||
| XM_005169837.4 | XP_005169894.1 | 3 - 4 | 2183 | 113 | ||
| GenBank mRNA | BC096906.1 | - | 6 - 7 | 2621 | 113 | |
More Information
Set Membership: |
|

ston2
Genomic Map