Assay Details
| Gene Symbol: ripor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045026.1 | NP_001038491.1 | 21 - 22 | 3129 | 97 | |
| XM_005159634.4 | XP_005159691.1 | 20 - 21 | 3029 | 97 | ||
| XM_009294297.3 | XP_009292572.1 | 20 - 21 | 2896 | 97 | ||
| XM_021468090.1 | XP_021323765.1 | 20 - 21 | 3144 | 97 | ||
| XM_021468091.1 | XP_021323766.1 | 20 - 21 | 3052 | 97 | ||
More Information
Set Membership: |
|

ripor2
Genomic Map