Assay Details
| Gene Symbol: cbx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199746.2 | NP_956040.2 | 2 - 3 | 318 | 73 | |
| GenBank mRNA | BC045443.1 | - | 2 - 3 | 304 | 73 | |
| BC075782.1 | - | 2 - 3 | 266 | 73 | ||
| BC165019.1 | - | 1 - 2 | 83 | 73 | ||
| EH457628.1 | - | 2 - 3 | 324 | 73 | ||
| EH545424.1 | - | 2 - 3 | 357 | 73 | ||
More Information
Set Membership: |
|

cbx1a
Genomic Map