Assay Details
| Gene Symbol: nr2f6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213239.1 | NP_998404.1 | 3 - 4 | 644 | 60 | |
| XM_005155689.4 | XP_005155746.1 | 3 - 4 | 717 | 60 | ||
| XM_017358215.2 | XP_017213704.1 | 4 - 5 | 850 | 60 | ||
| GenBank mRNA | BC065332.1 | - | 3 - 4 | 644 | 60 | |
| BC155820.1 | - | 3 - 4 | 691 | 60 | ||
More Information
Set Membership: |
|

nr2f6b
Genomic Map