Assay Details
| Gene Symbol: tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044919.1 | NP_001038384.1 | 2 - 3 | 533 | 75 | |
| GenBank mRNA | BC171428.1 | - | 2 - 3 | 478 | 75 | |
| BC171430.1 | - | 2 - 3 | 478 | 75 | ||
| CK146451.1 | - | 2 - 3 | 533 | 75 | ||
More Information
Set Membership: |
|

tmcc2
Genomic Map