Assay Details
| Gene Symbol: mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194361.2 | NP_919342.1 | 1 - 2 | 149 | 64 | |
| XM_017357587.2 | XP_017213076.1 | 1 - 2 | 181 | 64 | ||
| XM_017357591.2 | XP_017213080.1 | 1 - 2 | 180 | 64 | ||
| XM_017357594.2 | XP_017213083.1 | 1 - 2 | 178 | 64 | ||
| GenBank mRNA | AJ489219.1 | - | 1 - 2 | 151 | 64 | |
| BC085603.1 | - | 1 - 2 | 162 | 64 | ||
| BC164044.1 | - | 1 - 2 | 80 | 64 | ||
| CU458863.1 | - | 1 - 2 | 46 | 64 | ||
More Information
Set Membership: |
|

mpz
Genomic Map