Assay Details
| Gene Symbol: zgc:112294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020677.1 | NP_001018513.1 | 3 - 4 | 381 | 68 | |
| XM_017356090.2 | XP_017211579.1 | 3 - 4 | 446 | 68 | ||
| GenBank mRNA | BC095738.1 | - | 3 - 4 | 381 | 68 | |
More Information
Set Membership: |
|

zgc:112294
Genomic Map