Assay Details
| Gene Symbol: vamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200005.1 | NP_956299.1 | 3 - 4 | 485 | 141 | |
| GenBank mRNA | BC059626.1 | - | 3 - 4 | 485 | 141 | |
More Information
Set Membership: |
|

vamp2
Genomic Map