Assay Details
| Gene Symbol: gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207050.1 | NP_996933.1 | 5 - 6 | 693 | 63 | |
| GenBank mRNA | BC047197.1 | - | 5 - 6 | 589 | 63 | |
| BC066504.1 | - | 5 - 6 | 693 | 63 | ||
More Information
Set Membership: |
|

gmcl1
Genomic Map