Assay Details
| Gene Symbol: dgat1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002458.1 | NP_001002458.1 | 12 - 13 | 1256 | 101 | |
| XM_021466677.1 | XP_021322352.1 | 11 - 12 | 886 | 101 | ||
| GenBank mRNA | BC076012.1 | - | 12 - 13 | 1256 | 101 | |
More Information
Set Membership: |
|

dgat1b
Genomic Map