Assay Details
| Gene Symbol: dtnbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044814.2 | NP_001038279.2 | 1 - 2 | 239 | 66 | |
| XM_005170037.4 | XP_005170094.1 | 1 - 2 | 251 | 66 | ||
| GenBank mRNA | BC134087.1 | - | 1 - 2 | 239 | 66 | |
More Information
Set Membership: |
|

dtnbp1b
Genomic Map