Assay Details
| Gene Symbol: dtnbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044814.2 | NP_001038279.2 | 3 - 4 | 542 | 76 | |
| XM_005170037.4 | XP_005170094.1 | 3 - 4 | 545 | 76 | ||
| XM_009294215.3 | XP_009292490.1 | 2 - 3 | 837 | 76 | ||
| GenBank mRNA | BC134087.1 | - | 3 - 4 | 542 | 76 | |
More Information
Set Membership: |
|

dtnbp1b
Genomic Map