Assay Details
| Gene Symbol: egfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194424.1 | NP_919405.1 | 11 - 12 | 1455 | 75 | |
| GenBank mRNA | AY154658.1 | - | 11 - 12 | 1455 | 75 | |
| AY332223.1 | - | 11 - 12 | 1360 | 75 | ||
| AY533378.1 | - | 11 - 12 | 1360 | 75 | ||
| AY533379.1 | - | 11 - 12 | 1360 | 75 | ||
| BC163725.1 | - | 11 - 12 | 1326 | 75 | ||
More Information
Set Membership: |
|

egfra
Genomic Map