Assay Details
| Gene Symbol: atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199934.1 | NP_956228.1 | 1 - 2 | 142 | 64 | |
| GenBank mRNA | BC059611.1 | - | 1 - 2 | 142 | 64 | |
| BC152208.1 | - | 1 - 2 | 161 | 64 | ||
More Information
Set Membership: |
|

atp6v1g1
Genomic Map