Assay Details
| Gene Symbol: naa15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203321.2 | NP_976066.2 | 17 - 18 | 2323 | 71 | |
| GenBank mRNA | AY391464.1 | - | 17 - 18 | 2310 | 71 | |
More Information
Set Membership: |
|

naa15b
Genomic Map