Assay Details
| Gene Symbol: trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006029.1 | NP_001006029.1 | 4 - 5 | 548 | 63 | |
| GenBank mRNA | BC083391.1 | - | 4 - 5 | 548 | 63 | |
More Information
Set Membership: |
|

trappc6b
Genomic Map