Assay Details
| Gene Symbol: fbxo11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159668.1 | NP_001153140.1 | 14 - 15 | 1777 | 70 | |
| XM_005169554.4 | XP_005169611.1 | 14 - 15 | 2018 | 70 | ||
| GenBank mRNA | BC125857.1 | - | 14 - 15 | 70 | ||
More Information
Set Membership: |
|

fbxo11a
Genomic Map