Assay Details
| Gene Symbol: mbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131570.2 | NP_571645.2 | 3 - 4 | 370 | 72 | |
| XM_005171258.4 | XP_005171315.1 | 3 - 4 | 400 | 72 | ||
| GenBank mRNA | AF227738.1 | - | 3 - 4 | 353 | 72 | |
| BC095009.1 | - | 3 - 4 | 370 | 72 | ||
| BC165937.1 | - | 2 - 3 | 320 | 72 | ||
More Information
Set Membership: |
|

mbl2
Genomic Map