Assay Details
| Gene Symbol: mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152979.2 | NP_694511.1 | 2 - 3 | 265 | 62 | |
| XM_005155668.4 | XP_005155725.1 | 2 - 3 | 283 | 62 | ||
| GenBank mRNA | AF277172.1 | - | 2 - 3 | 214 | 62 | |
| BC045986.1 | - | 2 - 3 | 218 | 62 | ||
| BC065352.1 | - | 2 - 3 | 209 | 62 | ||
| CN321915.1 | - | 2 - 3 | 206 | 62 | ||
More Information
Set Membership: |
|

mkrn2
Genomic Map