Assay Details
| Gene Symbol: ghrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083872.1 | NP_001077341.1 | 1 | 46 | 63 | |
| XM_021476948.1 | XP_021332623.1 | 2 | 495 | 63 | ||
| GenBank mRNA | AM055940.1 | - | 1 | 46 | 63 | |
| EU908735.1 | - | 1 | 46 | 63 | ||
| GQ896512.1 | - | 2 | 99 | 63 | ||
More Information
Set Membership: |
|

ghrl
Genomic Map