Assay Details
| Gene Symbol: cdkn2aipnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006055.1 | NP_001006055.1 | 1 - 2 | 241 | 75 | |
| XM_005170293.3 | XP_005170350.1 | 1 - 2 | 286 | 75 | ||
| GenBank mRNA | BC083275.1 | - | 1 - 2 | 241 | 75 | |
More Information
Set Membership: |
|

cdkn2aipnl
Genomic Map