Assay Details
| Gene Symbol: ppm1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007313.1 | NP_001007314.1 | 3 - 4 | 1292 | 67 | |
| XM_005156264.4 | XP_005156321.1 | 3 - 4 | 1381 | 67 | ||
| GenBank mRNA | BC085660.1 | - | 3 - 4 | 1292 | 67 | |
| BC152616.1 | - | 3 - 4 | 1388 | 67 | ||
More Information
Set Membership: |
|

ppm1bb
Genomic Map