Assay Details
| Gene Symbol: angptl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126478.1 | NP_001119950.1 | 3 - 4 | 1437 | 63 | |
| XM_005167165.4 | XP_005167222.1 | 4 - 5 | 2004 | 63 | ||
| XM_005167166.4 | XP_005167223.1 | 4 - 5 | 2016 | 63 | ||
| GenBank mRNA | AB197678.1 | - | 3 - 4 | 1337 | 63 | |
| AB199648.1 | - | 3 - 4 | 1337 | 63 | ||
| BC163625.1 | - | 3 - 4 | 1444 | 63 | ||
| BC163635.1 | - | 3 - 4 | 1444 | 63 | ||
More Information
Set Membership: |
|

angptl2b
Genomic Map