Assay Details
| Gene Symbol: ndufa9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013459.2 | NP_001013477.2 | 10 - 11 | 1034 | 63 | |
| GenBank mRNA | BC068378.1 | - | 10 - 11 | 962 | 63 | |
| BC091545.1 | - | 10 - 11 | 983 | 63 | ||
| BC152139.1 | - | 10 - 11 | 984 | 63 | ||
| BC153422.1 | - | 10 - 11 | 1434 | 63 | ||
More Information
Set Membership: |
|

ndufa9a
Genomic Map