Assay Details
| Gene Symbol: rassf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002195.1 | NP_001002195.1 | 7 - 8 | 930 | 88 | |
| GenBank mRNA | BC074035.1 | - | 7 - 8 | 930 | 88 | |
| BC171489.1 | - | 7 - 8 | 831 | 88 | ||
| BC171493.1 | - | 7 - 8 | 831 | 88 | ||
More Information
Set Membership: |
|

rassf2b
Genomic Map