Assay Details
| Gene Symbol: maf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030239.1 | NP_001025410.1 | 3 - 4 | 698 | 91 | |
| XM_009303371.3 | XP_009301646.1 | 3 - 4 | 346 | 91 | ||
| GenBank mRNA | BC096891.1 | - | 3 - 4 | 698 | 91 | |
| BC165436.1 | - | 2 - 3 | 252 | 91 | ||
More Information
Set Membership: |
|

maf1b
Genomic Map