Assay Details
| Gene Symbol: maf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030239.1 | NP_001025410.1 | 7 - 8 | 1194 | 84 | |
| XM_009303371.3 | XP_009301646.1 | 7 - 8 | 842 | 84 | ||
| GenBank mRNA | BC096891.1 | - | 7 - 8 | 1194 | 84 | |
More Information
Set Membership: |
|

maf1b
Genomic Map