Assay Details
| Gene Symbol: kdm2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081692.2 | NP_001075161.2 | 3 - 4 | 462 | 101 | |
| XM_005160248.4 | XP_005160305.1 | 3 - 4 | 420 | 101 | ||
| XM_009293357.3 | XP_009291632.1 | 3 - 4 | 1048 | 101 | ||
| XM_009293362.3 | XP_009291637.1 | 3 - 4 | 419 | 101 | ||
| XM_009293365.3 | XP_009291640.1 | 3 - 4 | 349 | 101 | ||
| GenBank mRNA | BC129321.1 | - | 3 - 4 | 464 | 101 | |
More Information
Set Membership: |
|

kdm2aa
Genomic Map