Assay Details
| Gene Symbol: kdm2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081692.2 | NP_001075161.2 | 4 - 5 | 510 | 85 | |
| XM_005160248.4 | XP_005160305.1 | 4 - 5 | 468 | 85 | ||
| XM_009293357.3 | XP_009291632.1 | 4 - 5 | 1096 | 85 | ||
| XM_009293362.3 | XP_009291637.1 | 4 - 5 | 467 | 85 | ||
| XM_009293365.3 | XP_009291640.1 | 4 - 5 | 397 | 85 | ||
| GenBank mRNA | BC129321.1 | - | 4 - 5 | 512 | 85 | |
More Information
Set Membership: |
|

kdm2aa
Genomic Map