Assay Details
| Gene Symbol: ghra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083578.1 | NP_001077047.1 | 3 - 4 | 464 | 103 | |
| GenBank mRNA | BC134903.1 | - | 3 - 4 | 464 | 103 | |
| EU649774.1 | - | 3 - 4 | 556 | 103 | ||
More Information
Set Membership: |
|

ghra
Genomic Map