Assay Details
| Gene Symbol: snx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122855.2 | NP_001116327.1 | 1 - 2 | 2223 | 88 | |
| GenBank mRNA | BC169193.2 | - | 1 - 2 | 2229 | 88 | |
More Information
Set Membership: |
|

snx33
Genomic Map