Assay Details
| Gene Symbol: rhoab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212749.2 | NP_997914.2 | 4 - 5 | 459 | 60 | |
| XM_021478160.1 | XP_021333835.1 | 5 - 6 | 586 | 60 | ||
| GenBank mRNA | AY224600.1 | - | 3 - 4 | 406 | 60 | |
| AY865556.1 | - | 2 - 3 | 250 | 60 | ||
| BC075938.1 | - | 4 - 5 | 459 | 60 | ||
| BC152258.1 | - | 4 - 5 | 484 | 60 | ||
| BC171417.1 | - | 4 - 5 | 430 | 60 | ||
| BC171421.1 | - | 4 - 5 | 430 | 60 | ||
More Information
Set Membership: |
|

rhoab
Genomic Map