Assay Details
| Gene Symbol: MIR204B-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032990.1 | 1 | 41 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell migration inducing hyaluronidase 2 | CEMIP2 | TMEM2 | -788.3kb | Sense | Chr. 23 - 18599325 - 18671383 on NCBI Build EquCab3.0 |
| Downstream | Kruppel like factor 9 | KLF9 | 363.9kb | Sense | Chr. 23 - 19823693 - 19845722 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR204B-2
Genomic Map