Assay Details
| Gene Symbol: LOC100067436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497490.3 | XP_001497540.1 | 1211 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100067436| Gene Symbol: LOC100067436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497490.3 | XP_001497540.1 | 1211 | 97 | ||