Assay Details
| Gene Symbol: SERPINA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605427.3 | XP_005605484.1 | 797 | 97 | ||
| XM_005605428.3 | XP_005605485.1 | 736 | 97 | |||
More Information
Set Membership: |
|

SERPINA3
Genomic Map