Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918363.5 | XP_001918398.2 | 852 | 93 | ||
| GenBank mRNA | JL629584.1 | - | 2 - 3 | 307 | 93 | |
More Information
Set Membership: |
|

CHMP4A
Genomic Map