Assay Details
| Gene Symbol: KLHL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633521.1 | XP_023489289.1 | 709 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023633521.1 transcripts.
Set Membership: |
|

KLHL34
Genomic Map