Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646775.1 | XP_023502543.1 | 1278 | 62 | ||
| XM_023646779.1 | XP_023502547.1 | 1278 | 62 | |||
| GenBank mRNA | JL617528.1 | - | 2 - 3 | 61 | ||
More Information
Set Membership: |
|

ZNF248
Genomic Map