Assay Details
| Gene Symbol: COX6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492385.6 | XP_001492435.1 | 147 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001492385.6 transcripts.
Set Membership: |
|

COX6B1
Genomic Map