Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490394.6 | XP_001490444.3 | 360 | 104 | ||
| XM_014735780.2 | XP_014591266.1 | 360 | 104 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001490394.6, XM_014735780.2 transcripts.
Set Membership: |
|

HEXD
Genomic Map