Assay Details
| Gene Symbol: NPFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495079.4 | XP_001495129.1 | 818 | 55 | ||
| XM_023643707.1 | XP_023499475.1 | 664 | 55 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NPFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495079.4 | XP_001495129.1 | 818 | 55 | ||
| XM_023643707.1 | XP_023499475.1 | 664 | 55 | |||
Set Membership: |
|