Assay Details
| Gene Symbol: N6AMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278078.1 | NP_001265007.1 | 2 - 3 | 291 | 80 | |
| XM_025149445.1 | XP_025005213.1 | 2 - 3 | 303 | 80 | ||
| GenBank mRNA | BU252784.1 | - | 2 - 3 | 246 | 80 | |
| BX930346.2 | - | 2 - 3 | 290 | 80 | ||
| BX934446.1 | - | 2 - 3 | 285 | 80 | ||
| BX934970.1 | - | 2 - 3 | 284 | 80 | ||
More Information
Set Membership: |
|

N6AMT1
Genomic Map