Assay Details
| Gene Symbol: PAPOLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287288.2 | XP_015142774.1 | 19 - 20 | 2266 | 134 | |
| XM_015287291.2 | XP_015142777.1 | 19 - 20 | 2263 | 134 | ||
| GenBank mRNA | AF061758.1 | - | 15 - 16 | 1632 | 134 | |
More Information
Set Membership: |
|

PAPOLB
Genomic Map