Assay Details
| Gene Symbol: CXXC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293549.2 | XP_015149035.1 | 2 | 135 | 75 | |
| GenBank mRNA | CR353044.1 | - | 1 | 54 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015293549.2 transcripts.
Set Membership: |
|

CXXC5
Genomic Map