Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 14 - 15 | 1864 | 69 | |
| XM_003641367.4 | XP_003641415.1 | 16 - 17 | 2628 | 69 | ||
| XM_015286526.2 | XP_015142012.1 | 16 - 17 | 2854 | 69 | ||
| XM_015286527.2 | XP_015142013.1 | 17 - 18 | 2808 | 69 | ||
| XM_015286528.2 | XP_015142014.1 | 12 - 13 | 2194 | 69 | ||
| GenBank mRNA | AB046524.1 | - | 14 - 15 | 1864 | 69 | |
More Information
Set Membership: |
|

OVOA
Genomic Map