Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 15 - 16 | 1996 | 144 | |
| XM_003641367.4 | XP_003641415.1 | 17 - 18 | 2760 | 144 | ||
| XM_015286526.2 | XP_015142012.1 | 17 - 18 | 2986 | 144 | ||
| XM_015286527.2 | XP_015142013.1 | 18 - 19 | 2940 | 144 | ||
| XM_015286528.2 | XP_015142014.1 | 13 - 14 | 2326 | 144 | ||
| GenBank mRNA | AB046524.1 | - | 15 - 16 | 1996 | 144 | |
More Information
Set Membership: |
|

OVOA
Genomic Map