Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 37 - 38 | 5299 | 86 | |
| XM_003641367.4 | XP_003641415.1 | 39 - 40 | 6063 | 86 | ||
| XM_015286526.2 | XP_015142012.1 | 39 - 40 | 6289 | 86 | ||
| XM_015286527.2 | XP_015142013.1 | 40 - 41 | 6243 | 86 | ||
| XM_015286528.2 | XP_015142014.1 | 35 - 36 | 5629 | 86 | ||
| GenBank mRNA | AB046524.1 | - | 37 - 38 | 5299 | 86 | |
More Information
Set Membership: |
|

OVOA
Genomic Map