Assay Details
| Gene Symbol: GPR137B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031062.1 | NP_001026233.1 | 5 - 6 | 992 | 66 | |
| XM_015284054.2 | XP_015139540.1 | 6 - 7 | 1155 | 66 | ||
| GenBank mRNA | AJ719653.1 | - | 5 - 6 | 992 | 66 | |
More Information
Set Membership: |
|

GPR137B
Genomic Map