Assay Details
| Gene Symbol: AMY1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001528.2 | NP_001001528.2 | 2 - 3 | 394 | 87 | |
| GenBank mRNA | BX934793.1 | - | 2 - 3 | 394 | 87 | |
More Information
Set Membership: |
|

AMY1A
Genomic Map