Assay Details
| Gene Symbol: HCCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031275.1 | NP_001026446.1 | 2 - 3 | 351 | 78 | |
| XM_015290628.2 | XP_015146114.1 | 2 - 3 | 202 | 78 | ||
| GenBank mRNA | AJ851811.1 | - | 2 - 3 | 351 | 78 | |
| BX931817.1 | - | 2 - 3 | 226 | 78 | ||
| BX932410.2 | - | 2 - 3 | 235 | 78 | ||
More Information
Set Membership: |
|

HCCS
Genomic Map